Gene Transcripts

Gene Symbol
NCOA7
Entrez Gene ID
135112
Gene Name
nuclear receptor coactivator 7
Gene Aliases
ERAP140, ESNA1, NCOA7-AS, Nbla00052, Nbla10993, TLDC4, dJ187J11.3
Chromosome Location
Chr. 6: 125780948 - 125932030 on Build GRCh38
UniGene ID
Hs.171426
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001122842.2 NP_001116314.1 9 2225
NM_001199619.1 NP_001186548.1 10 2241
NM_001199620.1 NP_001186549.1 11 2359
NM_001199621.1 NP_001186550.1 8 2004
NM_181782.4 NP_861447.3 9 2258
XM_005266822.4 XP_005266879.1 9 2288
XM_006715340.3 XP_006715403.1 9 2215
XM_011535455.2 XP_011533757.1 9 2163
XM_017010269.1 XP_016865758.1 9 1974
XM_017010270.1 XP_016865759.1 10 2187
XM_017010271.1 XP_016865760.1 9 2185
XM_017010272.1 XP_016865761.1 9 2038
XM_017010273.1 XP_016865762.1 9 2255
XM_017010274.1 XP_016865763.1 9 2344
GenBank mRNA AB074156.1 - 8 1753
AF493978.1 - 10 2241
AJ420542.1 - 1 191
AK127512.1 - 9 2035
AK294558.1 - 8 1802
AK307395.1 - 10 2220
AL832621.2 - 9 2017
AL832628.2 - 9 1982
AL834442.1 - 7 1646
BC009399.2 - 1 922
BC033612.1 - 2 1096
BC137094.1 - 11 2275 9021091
BC137095.1 - 10 2157 9021092
BX537385.1 - 9 2053

View Full Product Details