Gene Transcripts

Gene Symbol
SSC4D
Entrez Gene ID
136853
Gene Name
scavenger receptor cysteine rich family member with 4 domains
Gene Aliases
S4D-SRCRB, SRCRB-S4D, SRCRB4D
Chromosome Location
Chr. 7: 76389329 - 76410025 on Build GRCh38
UniGene ID
Hs.567684
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080744.1 NP_542782.1 4 786
GenBank mRNA AJ312755.1 - 4 786
AK172783.1 - 4 744
BC113840.1 - 4 669

View Full Product Details