Gene Transcripts

Gene Symbol
LACC1
Entrez Gene ID
144811
Gene Name
laccase domain containing 1
Gene Aliases
C13orf31
Chromosome Location
Chr. 13: 43878834 - 43893932 on Build GRCh38
UniGene ID
Hs.210586
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128303.1 NP_001121775.1 5 1594
NM_153218.2 NP_694950.2 5 1430
XM_005266261.3 XP_005266318.1 5 2053
XM_006719766.3 XP_006719829.1 5 1211
XM_006719767.3 XP_006719830.1 5 1430
XM_011534933.2 XP_011533235.1 6 2248
XM_011534934.2 XP_011533236.1 5 1367
XM_017020394.1 XP_016875883.1 5 1487
GenBank mRNA AK096044.1 - 5 1430
BC035749.1 - 5 1399 5750179
BX538036.1 - 1 165

View Full Product Details