Gene Transcripts

Gene Symbol
ZDHHC8P1
Entrez Gene ID
150244
Gene Name
zinc finger DHHC-type containing 8 pseudogene 1
Gene Aliases
ZDHHC8P
Chromosome Location
Chr. 22: 23390605 - 23402612 on Build GRCh38
UniGene ID
Hs.386693
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003950.1 3 1108
GenBank mRNA AK056130.1 - 3 1041
AK092530.1 - 4 1182
AK092581.1 - 2 938
BC105936.1 - 1 690
BC105937.1 - 1 690
BC107600.1 - 1 690
CK902416.1 - 2 389

View Full Product Details