Gene Transcripts

Gene Symbol
C3orf67
Entrez Gene ID
200844
Gene Name
chromosome 3 open reading frame 67
Gene Aliases
Chromosome Location
Chr. 3: 58716415 - 59050078 on Build GRCh38
UniGene ID
Hs.368434
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198463.2 NP_940865.1 6 523
XM_005264929.2 XP_005264986.1 6 903
XM_006713013.3 XP_006713076.1 6 903
XM_011533451.2 XP_011531753.1 6 903
XM_011533453.2 XP_011531755.1 6 903
XM_011533455.2 XP_011531757.1 6 903
XM_011533456.1 XP_011531758.1 6 612
XM_011533457.1 XP_011531759.1 4 392
XM_011533458.2 XP_011531760.1 3 275
XM_011533459.2 XP_011531761.1 6 903
XM_011533460.2 XP_011531762.1 1 32
GenBank mRNA AK124111.1 - 6 525
AK296111.1 - 6 617
BC050317.1 - 6 890
BC132815.1 - 4 337 40146837
BC140002.1 - 7 633
BC151142.1 - 4 337 9020498

View Full Product Details