Gene Transcripts

Gene Symbol
DDIAS
Entrez Gene ID
220042
Gene Name
DNA damage induced apoptosis suppressor
Gene Aliases
C11orf82, noxin
Chromosome Location
Chr. 11: 82901695 - 82934659 on Build GRCh38
UniGene ID
Hs.165607
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145018.3 NP_659455.3 6 2816
XM_011544835.1 XP_011543137.1 5 2678
XM_011544836.2 XP_011543138.1 5 2633
XM_011544837.1 XP_011543139.1 4 2528
XM_011544838.2 XP_011543140.1 5 2662
XM_017017352.1 XP_016872841.1 4 2533
XM_017017353.1 XP_016872842.1 4 2500
GenBank mRNA AA477171.1 - 1 165
AK096157.1 - 6 2781
AK292399.1 - 6 2775
AK297366.1 - 4 2501
BC039268.1 - 6 2784 5296465

View Full Product Details