Gene Transcripts
- Gene Symbol
- PI16
- Entrez Gene ID
- 221476
- Gene Name
- peptidase inhibitor 16
- Gene Aliases
- CD364, CRISP9, MSMBBP, PSPBP
- Chromosome Location
- Chr. 6: 36948263 - 36964837 on Build GRCh38
- UniGene ID
- Hs.25391
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001199159.1 |
NP_001186088.1
|
2, 3 |
384 |
|
NM_153370.2 |
NP_699201.2
|
1, 2 |
489 |
|
XM_005248917.2 |
XP_005248974.1
|
1, 2 |
318 |
|
XM_011514375.2 |
XP_011512677.1
|
1, 2 |
318 |
|
XM_017010430.1 |
XP_016865919.1
|
1, 2 |
318 |
|
GenBank mRNA |
AK075470.1 |
- |
1, 2 |
184 |
|
AK124589.1 |
- |
4, 5 |
2008 |
|
AK303928.1 |
- |
1, 2 |
184 |
|
AK312785.1 |
- |
1, 2 |
182 |
|
AY358422.1 |
- |
1, 2 |
175 |
|
BC022399.1 |
- |
1, 2 |
183 |
4691115 |
BC035634.1 |
- |
1, 2 |
489 |
5217121 |
DA342875.1 |
- |
2, 3 |
348 |
|
DB178215.1 |
- |
2, 3 |
384 |
|
View Full Product Details