Gene Transcripts

Gene Symbol
C9orf43
Entrez Gene ID
257169
Gene Name
chromosome 9 open reading frame 43
Gene Aliases
Chromosome Location
Chr. 9: 113410644 - 113429684 on Build GRCh38
UniGene ID
Hs.632691
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278629.1 NP_001265558.1 2 448
NM_001278630.1 NP_001265559.1 2 305
NM_152786.2 NP_689999.1 2 521
XM_011518477.1 XP_011516779.1 2 448
XM_011518478.1 XP_011516780.1 2 448
GenBank mRNA AK301872.1 - 2 305
AK313867.1 - 2 437
BC026884.1 - 2 487 3455891
BC033037.2 - 2 406 4827930
DB048582.1 - 2 521
DB057586.1 - 2 449

View Full Product Details