Gene Transcripts

Gene Symbol
TAS2R43
Entrez Gene ID
259289
Gene Name
taste 2 receptor member 43
Gene Aliases
T2R43, T2R52
Chromosome Location
Chr. 12: 11091287 - 11092313 on Build GRCh38
UniGene ID
Hs.688195
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC101155.1 - 3 549

Gene Symbol
TAS2R45
Entrez Gene ID
259291
Gene Name
taste 2 receptor member 45
Gene Aliases
GPR59, T2R45, ZG24P
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.287378
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176886.2 NP_795367.2 3 549
GenBank mRNA AF264628.1 - 1 20
BC101154.1 - 3 549 40015678
BC101156.1 - 3 549 40015680

View Full Product Details