Gene Transcripts

Gene Symbol
B4GALNT3
Entrez Gene ID
283358
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 3
Gene Aliases
Chromosome Location
Chr. 12: 460377 - 561892 on Build GRCh38
UniGene ID
Hs.504416
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173593.3 NP_775864.3 15 2133
GenBank mRNA AB089940.1 - 15 2133
AK097681.1 - 2 518
AK131277.1 - 2 1524

View Full Product Details