Gene Transcripts

Gene Symbol
BTBD8
Entrez Gene ID
284697
Gene Name
BTB domain containing 8
Gene Aliases
Chromosome Location
Chr. 1: 92080305 - 92147840 on Build GRCh38
UniGene ID
Hs.676102
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183242.3 NP_899065.2 9 1326
GenBank mRNA AK296516.1 - 8 996
BC013922.1 - 9 1299 4048340

View Full Product Details