Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317948.1 | NP_001304877.1 | 3 | 508 | |
NM_173664.5 | NP_775935.1 | 3 | 508 | ||
XM_011534529.2 | XP_011532831.1 | 3 | 502 | ||
XM_011534530.2 | XP_011532832.1 | 3 | 502 | ||
XM_011534531.2 | XP_011532833.1 | 3 | 502 | ||
GenBank mRNA | BC059361.1 | - | 3 | 508 | |
DR001833.1 | - | 3 | 482 |