Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145017.1 | NP_001138489.1 | 11 | 1785 | |
NM_181774.3 | NP_861439.3 | 10 | 1662 | ||
XM_011537627.2 | XP_011535929.1 | 10 | 1584 | ||
XM_011537629.2 | XP_011535931.1 | 9 | 1492 | ||
XM_011537630.1 | XP_011535932.1 | 8 | 1009 | ||
XM_011537631.1 | XP_011535933.1 | 8 | 1020 | ||
XM_011537633.1 | XP_011535935.1 | 6 | 872 | ||
GenBank mRNA | AK127978.1 | - | 10 | 1655 | |
AK131483.1 | - | 9 | 3143 | ||
AY162215.1 | - | 10 | 1480 | ||
AY228365.1 | - | 3 | 289 | ||
BC101092.1 | - | 11 | 1508 | 40009824 | |
BC101093.1 | - | 11 | 1508 | 40009826 | |
BC101094.1 | - | 10 | 1385 | 40009832 | |
BC101095.2 | - | 10 | 1385 | 40009834 |