Gene Transcripts

Gene Symbol
BARHL2
Entrez Gene ID
343472
Gene Name
BarH like homeobox 2
Gene Aliases
Chromosome Location
Chr. 1: 90712022 - 90717237 on Build GRCh38
UniGene ID
Hs.451956
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020063.1 NP_064447.1 3 973
GenBank mRNA AY338397.1 - 3 973
BC126439.1 - 3 957 8992155
BC126441.1 - 3 957 8992157

View Full Product Details