Gene Transcripts

Gene Symbol
C12orf75
Entrez Gene ID
387882
Gene Name
chromosome 12 open reading frame 75
Gene Aliases
AGD3, OCC-1, OCC1
Chromosome Location
Chr. 12: 105330636 - 105371518 on Build GRCh38
UniGene ID
Hs.368938
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145199.1 NP_001138671.1 6 649
GenBank mRNA AB735446.1 - 6 589
AB735447.1 - 5 613
AJ311377.1 - 6 515
AJ311378.1 - 6 423
AK056999.1 - 6 521
BC013920.1 - 6 651 3838595
BC045812.1 - 6 613 4829271
BC061920.1 - 6 575 4413691
BC067897.1 - 6 623 5295847
BC126980.1 - 5 343 40108673
BC127012.1 - 5 343 40108672
BC131552.1 - 5 343

View Full Product Details