Gene Transcripts

Gene Symbol
OR14J1
Entrez Gene ID
442191
Gene Name
olfactory receptor family 14 subfamily J member 1
Gene Aliases
OR5U1, OR6-25, bA150A6.4, hs6M1-28
Chromosome Location
Chr. 6: 29306690 - 29307655 on Build GRCh38
UniGene ID
Hs.632004
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030946.1 NP_112208.1 1 326
GenBank mRNA BC136905.1 - 1 390

View Full Product Details