Gene Transcripts

Gene Symbol
OR10C1
Entrez Gene ID
442194
Gene Name
olfactory receptor family 10 subfamily C member 1 (gene/pseudogene)
Gene Aliases
OR10C1P, OR10C2, OR6-31, hs6M1-17
Chromosome Location
Chr. 6: 29439939 - 29440977 on Build GRCh38
UniGene ID
Hs.631997
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013941.3 NP_039229.3 1 965
GenBank mRNA BC136996.1 - 1 965
BC136997.1 - 1 965 9020989

View Full Product Details