Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001143.1 | NP_001134.1 | 5 | 587 | |
XM_017030042.1 | XP_016885531.1 | 6 | 658 | ||
GenBank mRNA | AY487421.1 | - | 5 | 561 | |
BC069138.1 | - | 5 | 587 | 7216997 | |
BC074976.1 | - | 5 | 548 | 30915513 | |
BC074977.1 | - | 5 | 548 | 30915296 | |
M86933.1 | - | 5 | 587 |