Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002786.3 | NP_002777.1 | 7 | 591 | |
NM_148976.2 | NP_683877.1 | 8 | 808 | ||
GenBank mRNA | AK290765.1 | - | 7 | 532 | |
BC002577.2 | - | 7 | 510 | 3140784 | |
BC005932.1 | - | 7 | 526 | 4078894 | |
BC008472.1 | - | 7 | 491 | 4282076 | |
BC009576.1 | - | 7 | 499 | 4105838 | |
BC015105.1 | - | 7 | 525 | 3950568 | |
BC015356.1 | - | 7 | 494 | 3452219 | |
BC022372.1 | - | 7 | 478 | 4770620 | |
BT006647.1 | - | 6 | 445 | ||
D00759.1 | - | 7 | 486 | ||
FJ224300.1 | - | 8 | 790 | ||
M64992.1 | - | 8 | 790 | ||
X61969.1 | - | 7 | 550 |