Gene Transcripts

Gene Symbol
TCAP
Entrez Gene ID
8557
Gene Name
titin-cap
Gene Aliases
CMD1N, CMH25, LGMD2G, T-cap, TELE, telethonin
Chromosome Location
Chr. 17: 39665346 - 39666554 on Build GRCh38
UniGene ID
Hs.77628
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003673.3 NP_003664.1 2 218
GenBank mRNA AJ000491.1 - 2 214
AK096328.1 - 2 1377
BC012628.1 - 2 219 4251080
BC013330.1 - 2 219 4273711
BF670957.1 - 2 219
DA899335.1 - 2 218
KU178429.1 - 2 204
KU178430.1 - 1 450

View Full Product Details