Gene Transcripts

Gene Symbol
CBX5
Entrez Gene ID
23468
Gene Name
chromobox 5
Gene Aliases
HEL25, HP1, HP1A
Chromosome Location
Chr. 12: 54230946 - 54280131 on Build GRCh38
UniGene ID
Hs.349283
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127321.1 NP_001120793.1 3 306
NM_001127322.1 NP_001120794.1 3 504
NM_012117.2 NP_036249.1 3 352
GenBank mRNA AK313506.1 - 3 313
BC006821.1 - 3 254 3448801
BP236173.1 - 3 306
BP336150.1 - 3 352
CR457418.1 - 2 186
DT217255.1 - 3 504
EU794599.1 - 3 321
L07515.1 - 3 321
S62077.1 - 3 320
U26311.1 - 2 184

View Full Product Details