Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_015362.4 | NP_056177.3 | 7 | 929 | |
NM_203413.2 | NP_981958.1 | 7 | 929 | ||
NM_203414.2 | NP_981959.1 | 6 | 1017 | ||
NM_203415.1 | NP_981960.1 | 7 | 822 | ||
XM_011523779.2 | XP_011522081.1 | 7 | 921 | ||
GenBank mRNA | AB013910.1 | - | 6 | 715 | |
AF087868.1 | - | 6 | 690 | ||
AF163262.1 | - | 6 | 648 | ||
AK225983.1 | - | 6 | 932 | ||
AK289940.1 | - | 6 | 932 | ||
AL713741.1 | - | 2 | 273 | ||
BC002762.2 | - | 7 | 894 | 3629369 | |
CA431021.1 | - | 3 | 498 | ||
CA450318.1 | - | 2 | 151 |