Gene Transcripts

Gene Symbol
ARRDC2
Entrez Gene ID
27106
Gene Name
arrestin domain containing 2
Gene Aliases
CLONE24945, PP2703
Chromosome Location
Chr. 19: 18001132 - 18014102 on Build GRCh38
UniGene ID
Hs.515249
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025604.2 NP_001020775.1 8 1373
NM_001286826.1 NP_001273755.1 8 1345
NM_015683.1 NP_056498.1 8 1348
GenBank mRNA AF131826.1 - 7 880
AF193051.1 - 8 1348
AK000689.1 - 8 1343
AK127940.1 - 8 1337
AK128588.1 - 2 2595
AK314657.1 - 8 1346
BC022516.1 - 8 1346 4817429

View Full Product Details