Gene Transcripts

Gene Symbol
POMP
Entrez Gene ID
51371
Gene Name
proteasome maturation protein
Gene Aliases
C13orf12, HSPC014, PNAS-110, UMP1
Chromosome Location
Chr. 13: 28659004 - 28678957 on Build GRCh38
UniGene ID
Hs.268742
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015932.5 NP_057016.1 5, 6 536
GenBank mRNA AF077200.1 - 5, 6 437
AF125097.1 - 5, 6 436
AF262975.1 - 5, 6 380
AF275807.1 - 5, 6 385
AK312118.1 - 5, 6 407
BC003390.1 - 5, 6 399 3446986
BC014334.1 - 5, 6 430 4045091
EF535527.1 - 5, 6 368
GU045555.1 - 5, 6 536
GU045556.1 - 5, 6 535

View Full Product Details