Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001170795.1 | NP_001164266.1 | 5 | 434 | |
NM_016085.4 | NP_057169.2 | 5 | 574 | ||
NM_080592.3 | NP_542159.3 | 5 | 754 | ||
GenBank mRNA | AB009017.1 | - | 5 | 447 | |
AF077037.1 | - | 5 | 436 | ||
AF144055.2 | - | 5 | 564 | ||
AF275744.1 | - | 6 | 428 | ||
AK291894.1 | - | 5 | 564 | ||
AY358968.1 | - | 5 | 427 | ||
BC002846.1 | - | 5 | 422 | ||
BC011006.1 | - | 5 | 428 | ||
BC021237.2 | - | 5 | 534 | 4303513 | |
BC035850.1 | - | 5 | 420 | 5753421 | |
BI194121.1 | - | 5 | 575 | ||
BQ420456.1 | - | 5 | 557 | ||
BQ718371.1 | - | 5 | 469 | ||
CF126865.1 | - | 5 | 567 |