Gene Transcripts

Gene Symbol
CALML5
Entrez Gene ID
51806
Gene Name
calmodulin like 5
Gene Aliases
CLSP
Chromosome Location
Chr. 10: 5498695 - 5499570 on Build GRCh38
UniGene ID
Hs.180142
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017422.4 NP_059118.2 1 160
GenBank mRNA AF172852.1 - 1 141
AK222744.1 - 1 160
BC039172.1 - 1 145 4753280

View Full Product Details