Gene Transcripts
- Gene Symbol
- FAM64A
- Entrez Gene ID
- 54478
- Gene Name
- family with sequence similarity 64 member A
- Gene Aliases
- CATS, RCS1
- Chromosome Location
- Chr. 17: 6444305 - 6451065 on Build GRCh38
- UniGene ID
- Hs.592116
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001195228.1 |
NP_001182157.1
|
2, 3 |
410 |
|
NM_019013.2 |
NP_061886.2
|
2, 3 |
410 |
|
XM_011523944.2 |
XP_011522246.1
|
2, 3 |
520 |
|
GenBank mRNA |
AK001018.1 |
- |
1, 2 |
350 |
|
AK001353.1 |
- |
2, 3 |
370 |
|
AK225613.1 |
- |
2, 3 |
372 |
|
BC005004.1 |
- |
2, 3 |
377 |
3533875 |
BC013966.2 |
- |
2, 3 |
356 |
2988318 |
KU178653.1 |
- |
1, 2 |
301 |
|
KU178654.1 |
- |
1, 2 |
301 |
|
View Full Product Details