Gene Transcripts

Gene Symbol
AVPI1
Entrez Gene ID
60370
Gene Name
arginine vasopressin induced 1
Gene Aliases
PP5395, VIP32, VIT32
Chromosome Location
Chr. 10: 97677422 - 97687432 on Build GRCh38
UniGene ID
Hs.23918
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021732.2 NP_068378.2 2 681
XM_017016494.1 XP_016871983.1 2 841
GenBank mRNA AF131791.1 - 2 681
AF218019.1 - 2 636
AF241786.1 - 2 683
AK024533.1 - 2 653
AK222493.1 - 2 654
AK223099.1 - 2 629
BC000877.1 - 2 292 3461724

View Full Product Details