Gene Transcripts

Gene Symbol
PAGR1
Entrez Gene ID
79447
Gene Name
PAXIP1 associated glutamate rich protein 1
Gene Aliases
C16orf53, GAS, PA1
Chromosome Location
Chr. 16: 29816207 - 29822495 on Build GRCh38
UniGene ID
Hs.702841
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024516.3 NP_078792.1 2 849
GenBank mRNA BC003640.2 - 2 545
EF195235.1 - 2 545

View Full Product Details