Gene Transcripts

Gene Symbol
LY6G5C
Entrez Gene ID
80741
Gene Name
lymphocyte antigen 6 complex, locus G5C
Gene Aliases
C6orf20, G5C, LY6G5CA, LY6G5CB, NG33
Chromosome Location
Chr. 6: 31676684 - 31680373 on Build GRCh38
UniGene ID
Hs.25738
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025262.3 NP_079538.3 3 374
GenBank mRNA AA179679.1 - 1 16
AA731226.1 - 1 361
AJ315540.1 - 4 478
AJ315541.1 - 4 598
AJ315542.1 - 3 365
AJ315543.1 - 3 459
AJ315544.1 - 2 1349

View Full Product Details