Gene Transcripts

Gene Symbol
C16orf92
Entrez Gene ID
146378
Gene Name
chromosome 16 open reading frame 92
Gene Aliases
Chromosome Location
Chr. 16: 30023334 - 30024702 on Build GRCh38
UniGene ID
Hs.651588
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109659.1 NP_001103129.1 2 120
NM_001109660.1 NP_001103130.1 2 200
GenBank mRNA AK058133.1 - 2 200
BC101433.1 - 2 120 40021839
BC101434.1 - 2 120 40021838
BC101435.1 - 2 120 40021831
BC101436.1 - 2 200 40021833
BC127955.1 - 2 200 40023544

View Full Product Details