Gene Transcripts
- Gene Symbol
- SPDYC
- Entrez Gene ID
- 387778
- Gene Name
- speedy/RINGO cell cycle regulator family member C
- Gene Aliases
- RINGOC, Ringo2
- Chromosome Location
- Chr. 11: 65170236 - 65173666 on Build GRCh38
- UniGene ID
- Hs.450642
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001008778.1 |
NP_001008778.1
|
4, 5 |
333 |
|
XM_017017705.1 |
XP_016873194.1
|
3, 4 |
727 |
|
XM_017017706.1 |
XP_016873195.1
|
3, 4 |
727 |
|
XM_017017707.1 |
XP_016873196.1
|
3, 4 |
727 |
|
XM_017017708.1 |
XP_016873197.1
|
3, 4 |
727 |
|
XM_017017709.1 |
XP_016873198.1
|
3, 4 |
727 |
|
XM_017017710.1 |
XP_016873199.1
|
3, 4 |
727 |
|
GenBank mRNA |
AY820305.1 |
- |
4, 5 |
333 |
|
BC137244.1 |
- |
4, 5 |
415 |
9021241 |
BC137246.1 |
- |
4, 5 |
415 |
9021243 |
BC144548.1 |
- |
4, 5 |
415 |
9053075 |
View Full Product Details