Gene Transcripts

Gene Symbol
ATXN7L3B
Entrez Gene ID
552889
Gene Name
ataxin 7 like 3B
Gene Aliases
lnc-SCA7
Chromosome Location
Chr. 12: 74537771 - 74541452 on Build GRCh38
UniGene ID
Hs.744849
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001136262.1 NP_001129734.1 1 652
GenBank mRNA AK056642.1 - 1 596
AL713650.2 - 1 545
BC009111.1 - 1 556 3460701
BX647460.2 - 1 545

View Full Product Details