Gene Transcripts

Gene Symbol
C15orf52
Entrez Gene ID
388115
Gene Name
chromosome 15 open reading frame 52
Gene Aliases
Chromosome Location
Chr. 15: 40331452 - 40340967 on Build GRCh38
UniGene ID
Hs.32433
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207380.2 NP_997263.2 11 1487
GenBank mRNA AK125329.1 - 11 1480
AK126035.1 - 7 877
BC140897.1 - 7 876 9021765
BC140902.1 - 7 876 9021770
CR749408.1 - 8 1181

View Full Product Details