Gene Transcripts

Gene Symbol
UBE2NL
Entrez Gene ID
389898
Gene Name
ubiquitin conjugating enzyme E2 N like (gene/pseudogene)
Gene Aliases
Li174
Chromosome Location
Chr. X: 143884071 - 143885255 on Build GRCh38
UniGene ID
Hs.585177
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012989.2 NP_001013007.1 1 477
NR_121210.1 1 477
GenBank mRNA GU727624.1 - 1 477

View Full Product Details