Gene Transcripts

Gene Symbol
ARL5C
Entrez Gene ID
390790
Gene Name
ADP ribosylation factor like GTPase 5C
Gene Aliases
ARL12
Chromosome Location
Chr. 17: 39156894 - 39166161 on Build GRCh38
UniGene ID
Hs.568007
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001143968.1 NP_001137440.1 5 849

View Full Product Details