Gene Transcripts

Gene Symbol
SMU1
Entrez Gene ID
55234
Gene Name
DNA replication regulator and spliceosomal factor
Gene Aliases
BWD, SMU-1, fSAP57
Chromosome Location
Chr. 9: 33041852 - 33076716 on Build GRCh38
UniGene ID
Hs.655351
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018225.2 NP_060695.2 12 4787
XM_005251503.4 XP_005251560.1 11 4623
GenBank mRNA AL512690.1 - 1 645

View Full Product Details