Gene Transcripts
- Gene Symbol
- AGAP9
- Entrez Gene ID
- 642517
- Gene Name
- ArfGAP with GTPase domain, ankyrin repeat and PH domain 9
- Gene Aliases
- AGAP-9, CTGLF6, bA301J7.2
- Chromosome Location
- Chr. 10: 47501854 - 47523638 on Build GRCh38
- UniGene ID
- Hs.645346
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK096908.1 |
- |
1 |
650 |
|
View Full Product Details