Gene Transcripts

Gene Symbol
C22orf34
Entrez Gene ID
348645
Gene Name
chromosome 22 open reading frame 34
Gene Aliases
Chromosome Location
Chr. 22: 49414524 - 49657542 on Build GRCh38
UniGene ID
Hs.133159
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017028796.1 XP_016884285.1 7 10337
GenBank mRNA AK128192.1 - 2 628

View Full Product Details