Gene Transcripts

Gene Symbol
C8orf58
Entrez Gene ID
541565
Gene Name
chromosome 8 open reading frame 58
Gene Aliases
Chromosome Location
Chr. 8: 22599601 - 22604150 on Build GRCh38
UniGene ID
Hs.743508
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013842.2 NP_001013864.1 6 1026
GenBank mRNA BC012750.2 - 5 817

View Full Product Details