Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006544.3 | NP_006535.1 | 10 | 1129 | |
XM_005267272.3 | XP_005267329.1 | 10 | 1214 | ||
GenBank mRNA | AK295085.1 | - | 10 | 1036 | |
AK303531.1 | - | 9 | 872 | ||
AK312523.1 | - | 10 | 1134 | ||
BC041126.1 | - | 10 | 1095 | 3914527 | |
U85946.1 | - | 10 | 1073 |