Gene Transcripts

Gene Symbol
EHD1
Entrez Gene ID
10938
Gene Name
EH domain containing 1
Gene Aliases
H-PAST, HPAST1, PAST, PAST1
Chromosome Location
Chr. 11: 64852727 - 64879713 on Build GRCh38
UniGene ID
Hs.523774
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282444.1 NP_001269373.1 7 1886
NM_001282445.1 NP_001269374.1 6 1564
NM_006795.3 NP_006786.2 5 1743
XM_011544739.1 XP_011543041.1 6 1561
GenBank mRNA AF001434.1 - 5 1694
AK096747.1 - 1 597
AK097842.1 - 7 1851
AK098723.1 - 5 948
AK124613.1 - 4 2661
AK126581.1 - 3 1321
AK297654.1 - 6 1422
AK299096.1 - 6 1475
AK300326.1 - 6 1524
AK312316.1 - 5 1596
AY007161.1 - 5 1490
BC104799.1 - 5 1635 8143802
BC104825.1 - 5 1635 8143828

View Full Product Details