Gene Transcripts
- Gene Symbol
- FAM71E1
- Entrez Gene ID
- 112703
- Gene Name
- family with sequence similarity 71 member E1
- Gene Aliases
- Chromosome Location
- Chr. 19: 50466643 - 50476746 on Build GRCh38
- UniGene ID
- Hs.448941
- Species
- Homo sapiens
- Gene Symbol
- EMC10
- Entrez Gene ID
- 284361
- Gene Name
- ER membrane protein complex subunit 10
- Gene Aliases
- C19orf63, HSM1, HSS1
- Chromosome Location
- Chr. 19: 50476477 - 50505905 on Build GRCh38
- UniGene ID
- Hs.448941
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AY194293.2 |
- |
2 |
270 |
|
View Full Product Details