Gene Transcripts

Gene Symbol
SOWAHA
Entrez Gene ID
134548
Gene Name
sosondowah ankyrin repeat domain family member A
Gene Aliases
ANKRD43
Chromosome Location
Chr. 5: 132813324 - 132816797 on Build GRCh38
UniGene ID
Hs.13308
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175873.5 NP_787069.4 1 1661
GenBank mRNA AK090823.1 - 1 88
AK092782.1 - 1 604
BC104771.1 - 1 790 8143774
BC104775.1 - 1 790 8143778

View Full Product Details