Gene Transcripts

Gene Symbol
CCDC12
Entrez Gene ID
151903
Gene Name
coiled-coil domain containing 12
Gene Aliases
Chromosome Location
Chr. 3: 46917905 - 46982031 on Build GRCh38
UniGene ID
Hs.631918
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277074.1 NP_001264003.1 3 224
NM_144716.5 NP_653317.2 4 379
NR_102269.1 3 242
XM_011533390.2 XP_011531692.1 3 319
XM_011533391.2 XP_011531693.1 4 385
XM_011533392.2 XP_011531694.1 4 337
XM_011533393.2 XP_011531695.1 2 1025
XM_011533394.2 XP_011531696.1 3 861
XM_011533396.2 XP_011531698.1 3 578
GenBank mRNA AK096749.1 - 3 202
AK295532.1 - 3 202
AK303160.1 - 3 216
BC020830.1 - 3 205 4771328
BM676030.1 - 5 653
BM803652.1 - 3 149
CN430023.1 - 4 334
CV574708.1 - 5 455
DA952452.1 - 242
DC379144.1 - 3 227

View Full Product Details