Gene Transcripts

Gene Symbol
MDGA2
Entrez Gene ID
161357
Gene Name
MAM domain containing glycosylphosphatidylinositol anchor 2
Gene Aliases
MAMDC1, c14_5286
Chromosome Location
Chr. 14: 46836518 - 47675319 on Build GRCh38
UniGene ID
Hs.436380
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113498.2 NP_001106970.3 11 2489
NM_182830.4 NP_878250.2 11 2195
XM_011536519.2 XP_011534821.1 11 2344
XM_011536525.2 XP_011534827.1 8 1626
XM_017021060.1 XP_016876549.1 7 1455
GenBank mRNA AK123307.1 - 1 60
AK309211.1 - 11 2492
AY328482.1 - 5 1122
AY358125.1 - 10 2069
AY369208.1 - 11 2285
BC142947.1 - 11 2212

View Full Product Details