Gene Transcripts

Gene Symbol
ABHD3
Entrez Gene ID
171586
Gene Name
abhydrolase domain containing 3
Gene Aliases
LABH3
Chromosome Location
Chr. 18: 21650897 - 21704805 on Build GRCh38
UniGene ID
Hs.397978
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308256.1 NP_001295185.1 3 505
NM_138340.4 NP_612213.2 3 505
XM_011525847.2 XP_011524149.1 3 478
XM_017025571.1 XP_016881060.1 3 481
XM_017025572.1 XP_016881061.1 3 355
XM_017025573.1 XP_016881062.1 3 265
XM_017025574.1 XP_016881063.1 1 37
GenBank mRNA AF007152.1 - 3 464
AK024880.1 - 3 379
AK298728.1 - 3 404
BC021196.2 - 3 465 3942164
BI918326.1 - 3 457
DB126083.1 - 3 505

View Full Product Details