Gene Transcripts
- Gene Symbol
- ELF3
- Entrez Gene ID
- 1999
- Gene Name
- E74 like ETS transcription factor 3
- Gene Aliases
- EPR-1, ERT, ESE-1, ESX
- Chromosome Location
- Chr. 1: 202010519 - 202017188 on Build GRCh38
- UniGene ID
- Hs.603657
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001114309.1 |
NP_001107781.1
|
2, 3 |
431 |
|
NM_004433.4 |
NP_004424.3
|
2, 3 |
317 |
|
XM_005244942.2 |
XP_005244999.1
|
2, 3 |
360 |
|
GenBank mRNA |
AF017307.1 |
- |
2, 3 |
366 |
|
AF517841.1 |
- |
2, 3 |
299 |
|
AK225446.1 |
- |
2, 3 |
254 |
|
AK310814.1 |
- |
2, 3 |
309 |
|
AK312273.1 |
- |
2, 3 |
293 |
|
BC003569.1 |
- |
2, 3 |
285 |
3537846 |
BX537368.1 |
- |
1, 2 |
3354 |
|
CR457106.1 |
- |
1, 2 |
164 |
|
CV570991.1 |
- |
2, 3 |
317 |
|
DA591406.1 |
- |
2, 3 |
407 |
|
U66894.1 |
- |
2, 3 |
259 |
|
U73843.1 |
- |
2, 3 |
283 |
|
U73844.1 |
- |
2, 3 |
283 |
|
U97156.1 |
- |
2, 3 |
288 |
|
View Full Product Details