Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001114309.1 | NP_001107781.1 | 3 | 576 | |
NM_004433.4 | NP_004424.3 | 3 | 462 | ||
XM_005244942.2 | XP_005244999.1 | 3 | 505 | ||
GenBank mRNA | AF016295.1 | - | 3 | 423 | |
AF017307.1 | - | 3 | 511 | ||
AF517841.1 | - | 3 | 444 | ||
AK225446.1 | - | 3 | 399 | ||
AK309265.1 | - | 2 | 270 | ||
AK310814.1 | - | 3 | 454 | ||
AK312273.1 | - | 3 | 438 | ||
BC003569.1 | - | 3 | 430 | 3537846 | |
BX537368.1 | - | 2 | 3499 | ||
CR457106.1 | - | 2 | 309 | ||
CV570991.1 | - | 3 | 462 | ||
DA591406.1 | - | 3 | 552 | ||
U66894.1 | - | 3 | 404 | ||
U73843.1 | - | 3 | 428 | ||
U73844.1 | - | 3 | 428 | ||
U97156.1 | - | 3 | 433 |