Gene Transcripts

Gene Symbol
CEP112
Entrez Gene ID
201134
Gene Name
centrosomal protein 112
Gene Aliases
CCDC46, MACOCO
Chromosome Location
Chr. 17: 65635540 - 66192131 on Build GRCh38
UniGene ID
Hs.408676
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037325.2 NP_001032402.1 5 755
NM_001199165.2 NP_001186094.1 25 2886
NM_001302891.1 NP_001289820.1 23 2836
NR_126542.1 4 585
XM_005257119.4 XP_005257176.1 25 2980
XM_005257125.3 XP_005257182.1 17 1970
XM_005257126.4 XP_005257183.1 14 1635
XM_006721740.2 XP_006721803.1 25 2852
XM_011524461.2 XP_011522763.1 25 2993
XM_011524464.2 XP_011522766.1 23 2769
XM_011524465.2 XP_011522767.1 22 2309
XM_011524466.2 XP_011522768.1 15 1788
XM_011524467.2 XP_011522769.1 14 1630
XM_017024302.1 XP_016879791.1 23 2723
GenBank mRNA AA781351.1 - 3 279
AF458591.1 - 15 1793
AK098070.1 - 15 1705
AK299075.1 - 25 2837
BC029524.1 - 5 755
BC131805.1 - 23 2664 40119069
BG203878.1 - 52
BG531260.1 - 2 73
BX097629.1 - 5 433

View Full Product Details